Cadmium chloride, ultra dry, 99.998% (metals basis)
Cadmium chloride, ultra dry, 99.998% (metals basis)
Cadmium chloride, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals

Cadmium chloride, ultra dry, 99.998% (metals basis)

CAS: 10108-64-2 | CdCl2 | 183.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013667.06
also known as 013667-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS10108-64-2
IUPAC Namecadmium(2+) dichloride
Molecular FormulaCdCl2
InChI KeyYKYOUMDCQGMQQO-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Cd++]
View more
SpecificationsSpecification SheetSpecification Sheet
Residual water< 100 ppm
Total Metal Impurities0.002% max.
Particle Size-10 Mesh
Appearance (Color)White
FormBeads
Cadmium chloride is utilized in the preparation of CdTe nanocrystals, highly luminescent. It is also employed in analytical chemistry, photography, dyeing and calico printing, ingredient of electroplating baths, manufacture of special mirrors, the preparation of cadmium standard, analysis of sulfides, and testing pyridine bases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only