Potassium chloride, ultra dry, 99.998% (metals basis)
Potassium chloride, ultra dry, 99.998% (metals basis)
Potassium chloride, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals

Potassium chloride, ultra dry, 99.998% (metals basis)

CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013682.18
also known as 013682-18
Price (USD)
502.65
Online Exclusive
558.00
Save 55.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
502.65
Online Exclusive
558.00
Save 55.35 (10%)
Each
Chemical Identifiers
CAS7447-40-7
IUPAC Namepotassium chloride
Molecular FormulaClK
InChI KeyWCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES[Cl-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.002% max.
Particle Size-10 Mesh
The main use of potassium chloride is in the manufacture of fertilizers. It finds use in medicine as an electrolyte replenisher and to treat hypokalemia (a condition in which the level of potassium will be low in blood) with its pharmaceutical preparation. Furthermore, it finds use as a flux agent in welding and metal casting. It is useful in water softener units, certain de-icing products and calibration of the radiation monitoring equipment. It finds use as a scotophor in dark-trace CRTs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only