Sodium chloride, ultra dry, 99.998% (metals basis)
Sodium chloride, ultra dry, 99.998% (metals basis)
Sodium chloride, ultra dry, 99.998% (metals basis)
Thermo Scientific Chemicals

Sodium chloride, ultra dry, 99.998% (metals basis)

CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 013683.14
also known as 013683-14
Price (USD)
456.65
Special offer
Online exclusive
Ends: 30-Jun-2026
537.00
Save 80.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
456.65
Special offer
Online exclusive
Ends: 30-Jun-2026
537.00
Save 80.35 (15%)
Each
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.002% max.
Particle Size-10 Mesh
Sodium Chloride is widely used in biochemistry and molecular biology applications. It is used in a wide variety of biochemical applications including creating density gradients, regulating biological functions and as a diluent to increase ionic strength in buffers or culture media, a component of PBS and SSC buffers. It has also been used for precipitation of DNA from SDS-containing samples and in the removal of small nucleic acid fragments from plasmid DNA preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only