Scandium(III) iodide, ultra dry, 99.999% (REO)
Scandium(III) iodide, ultra dry, 99.999% (REO)
Scandium(III) iodide, ultra dry, 99.999% (REO)
Scandium(III) iodide, ultra dry, 99.999% (REO)
Thermo Scientific Chemicals

Scandium(III) iodide, ultra dry, 99.999% (REO)

CAS: 14474-33-0 | I3Sc | 425.669 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 013687.03
also known as 013687-03
Price (USD)
266.65
Special offer
Online exclusive
Ends: 30-Jun-2026
314.00
Save 47.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
266.65
Special offer
Online exclusive
Ends: 30-Jun-2026
314.00
Save 47.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS14474-33-0
IUPAC Namescandium(3+) triiodide
Molecular FormulaI3Sc
InChI KeyHUIHCQPFSRNMNM-UHFFFAOYSA-K
SMILES[Sc+3].[I-].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Residual water< 100 ppm
Total Rare Earth Oxide Impurities0.001% max.
Scandium iodide is used in high-intensity lighting.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only