Zinc chloride, ultra dry, 99.99% (metals basis)
Zinc chloride, ultra dry, 99.99% (metals basis)
Zinc chloride, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals

Zinc chloride, ultra dry, 99.99% (metals basis)

CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013694.18
also known as 013694-18
Price (USD)
296.65
Special offer
Online exclusive
Ends: 30-Jun-2026
349.00
Save 52.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
296.65
Special offer
Online exclusive
Ends: 30-Jun-2026
349.00
Save 52.35 (15%)
Each
Chemical Identifiers
CAS7646-85-7
IUPAC Namezinc(2+) dichloride
Molecular FormulaCl2Zn
InChI KeyJIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Zn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Particle Size-10 Mesh
FormBeads
Residual water< 100 ppm
Total Metal Impurities0.01% max.
Appearance (Color)White
Zinc(II) chloride is used as a catalyst, as a dehydrating and condensing agent, soldering flux and metal etchant. It is also employed in preserving anatomical specimens, wood preservatives, deodorant, disinfecting and embalming materials. It is employed as a mordant in printing and dyeing materials, and in the vulcanizing processes of fiber and rubber. It is useful as an electrolyte in dry cell batteries, in metal industry, in galvanizing iron and as an electrolyte for electroplating. It is useful in an organic synthesis, for example in the Friedel-crafts acylation, Fisher indole synthesis, Lucas reagent (zinc chloride with HCl), activation of allylic/benzylic halides towards reaction with olefins or with sodium cyanoborohydride catalyzed reduction of halides, organozinc reagents for use in Negishi coupling, and aldol condensation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only