Hexaamminecobalt(III) chloride, 97%
Hexaamminecobalt(III) chloride, 97%
Hexaamminecobalt(III) chloride, 97%
Thermo Scientific Chemicals

Hexaamminecobalt(III) chloride, 97%

CAS: 10534-89-1 | [Co(NH3)6]Cl3
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013769.14
also known as 013769-14
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
101.65
Online Exclusive
113.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS10534-89-1
IUPAC Namecobalt(3+) hexaamine trichloride
Molecular FormulaCl3CoH18N6
InChI KeyJXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILESN.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)96.1% min
Hexaamminecobalt(III) chloride is used in X-ray crystallography and NMR. It is useful in DNA condensation studies. It is suitable for induction of transitions of DNA from B to A or Z forms, induction of nucleic acid crystal growth, stabilization of tertiary tRNA interactions, preparation of ligation buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only