Stainless Steel flake, -325 mesh, 0.4-1.2 micron thick, Type 316-L
Thermo Scientific Chemicals

Stainless Steel flake, -325 mesh, 0.4-1.2 micron thick, Type 316-L

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 013787.30
also known as 013787-30
Price (USD)
58.00
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
58.00
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
Chemical Name or MaterialStainless Steel flake, Type 316-L
MDL NumberMFCD00148499
Mesh Size−325 Mesh
Particle Size0.4 to 1.2 Micron thick
View more
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only