Nickel spheres, silver coated, -250 mesh, Ni typically 99.8% (metals basis)
Nickel spheres, silver coated, -250 mesh, Ni typically 99.8% (metals basis)
Nickel spheres, silver coated, -250 mesh, Ni typically 99.8% (metals basis)
Thermo Scientific Chemicals

Nickel spheres, silver coated, -250 mesh, Ni typically 99.8% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 013793.14
also known as 013793-14
Price (USD)
60.65
Online Exclusive
67.20
Save 6.55 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
67.20
Save 6.55 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
Avg. Mol. Wt. or Mol. Wt. RangeNi:Ag; 85:15 wt%
CAS7440-02-0
ChEBICHEBI:28112
Chemical Name or MaterialNickel spheres
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only