Sodium hydrogen sulfate monohydrate, Reagent Grade
Sodium hydrogen sulfate monohydrate, Reagent Grade
Sodium hydrogen sulfate monohydrate, Reagent Grade
Thermo Scientific Chemicals

Sodium hydrogen sulfate monohydrate, Reagent Grade

CAS: 10034-88-5 | H3NaO5S | 138.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Catalog number 013799.36
also known as 013799-36
Price (USD)
87.65
Online Exclusive
96.80
Save 9.15 (9%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
87.65
Online Exclusive
96.80
Save 9.15 (9%)
Each
Chemical Identifiers
CAS10034-88-5
IUPAC Namesodium hydrate hydrogen sulfate
Molecular FormulaH3NaO5S
InChI KeyJXHZRQHZVYDRGX-UHFFFAOYSA-M
SMILESO.[Na+].OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
FormCrystals, moist in appearance
Free acid (titration)Acidity (as H₂SO₄) 35.0-36.5%
Impurity contentAs:≤1 ppm
Impurity contentCl: ≤20 ppm
View more
Sodium hydrogen sulfate monohydrate is used in metal finishing and cleaning products. It is used to lower the pH of water for an effective chlorination in swimming pools. It is also used as a urine acidifier to reduce urinary stones in cats. Further, it is used in controlling outbreaks of crown-of-thorns starfish. It is a primary ingredient used in jewelry making to clean the surfaces. In addition to this, it is used as a food additive and serves as a reagent for the decomposition of basic metal oxides by fusion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only