Aluminum ammonium sulfate dodecahydrate, Reagent Grade
Aluminum ammonium sulfate dodecahydrate, Reagent Grade
Aluminum ammonium sulfate dodecahydrate, Reagent Grade
Thermo Scientific Chemicals

Aluminum ammonium sulfate dodecahydrate, Reagent Grade

CAS: 7784-26-1 | AlH28NO20S2 | 453.313 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 013802.36
also known as 013802-36
Price (USD)
93.70
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
93.70
Each
Chemical Identifiers
CAS7784-26-1
IUPAC Namealuminium(3+) ammonium dodecahydrate disulfate
Molecular FormulaAlH28NO20S2
InChI KeyWZUKKIPWIPZMAS-UHFFFAOYSA-K
SMILES[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Formpowder or crystals
Assay≥98.0%
Assay (unspecified)≥98.0%
Water and sewage purification, dyeing and printing fabrics, manufacturing of pigments, paper, artificial gems, cements, in electrolytic copper platingAluminum ammonium sulfate dodecahydrate acts as a crystalline double sulfate. It is used in water purification, in vegetable glues, in porcelain cements, in deodorant, in tanning and in fireproofing textiles. It serves as an ingredient in animal repellant sprays. Further, it is used in the preparation of Alum (Al(OH)3) gel. In addition, it is used in cosmetic formulation, fertilizers, food preservation and in the heating system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only