Cesium, plasma standard solution, Specpure™ Cs 1000μg/mL
Cesium, plasma standard solution, Specpure™ Cs 1000μg/mL
Cesium, plasma standard solution, Specpure™ Cs 1000μg/mL
Thermo Scientific Chemicals

Cesium, plasma standard solution, Specpure™ Cs 1000μg/mL

CAS: 534-17-8 | CCs2O3 | 325.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
500 mL
Catalog number 013825.AD
also known as 013825-AD
Price (USD)
56.70
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
56.70
Each
Specifications
CAS534-17-8
CAS7697-37-2
CAS7732-18-5
Chemical Name or MaterialCesium
ColorColorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only