Niobium, plasma standard solution, Specpure™ Nb 1000μg/mL, Thermo Scientific Chemicals
Niobium, plasma standard solution, Specpure™ Nb 1000μg/mL, Thermo Scientific Chemicals
Niobium, plasma standard solution, Specpure™ Nb 1000μg/mL, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium, plasma standard solution, Specpure™ Nb 1000μg/mL, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013831.AE
also known as 013831-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
CAS3-1-7440
ChEBICHEBI:33344
Chemical Name or MaterialNiobium
DOT InformationHazard Class: 8; Packaging Group: III
Health Hazard 1H290-H311-H315-H319-H332
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only