Arsenic, plasma standard solution, Specpure™ As 1000μg/mL
Arsenic, plasma standard solution, Specpure™ As 1000μg/mL
Arsenic, plasma standard solution, Specpure™ As 1000μg/mL
Thermo Scientific Chemicals

Arsenic, plasma standard solution, Specpure™ As 1000μg/mL

CAS: 7440-38-2 | As | 74.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013836.AD
also known as 013836-AD
Price (USD)
-
Quantity:
50 mL
Request bulk or custom format
Specifications
CAS7440-38-2
CAS7732-18-5
ChEBICHEBI:27563
Chemical Name or MaterialArsenic
ColorColorless
View more
Arsenic, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only