Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL
Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL
Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL
Thermo Scientific Chemicals

Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013839.AD
also known as 013839-AD
Price (USD)
57.40
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
57.40
Each
Specifications
Chemical Name or MaterialNickel, plasma standard solution, Specpure™
DOT InformationHazard Class: 8; Packaging Group: II
Health Hazard 1H290-H314-H335-H350i-H360D
Health Hazard 2GHS H Statement
H314-H317-H208
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only