Germanium, plasma standard solution, Specpure™ Ge 1000μg/mL
Germanium, plasma standard solution, Specpure™ Ge 1000μg/mL
Germanium, plasma standard solution, Specpure™ Ge 1000μg/mL
Thermo Scientific Chemicals

Germanium, plasma standard solution, Specpure™ Ge 1000μg/mL

CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013841.AD
also known as 013841-AD
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.10
Save 8.45 (14%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.10
Save 8.45 (14%)
Each
Specifications
CAS7440-56-4
CAS7664-39-3
CAS7697-37-2
CAS7732-18-5
ChEBICHEBI:30441
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only