Sulfur, plasma standard solution, Specpure™ S 1000μg/mL
Sulfur, plasma standard solution, Specpure™ S 1000μg/mL
Sulfur, plasma standard solution, Specpure™ S 1000μg/mL
Thermo Scientific Chemicals

Sulfur, plasma standard solution, Specpure™ S 1000μg/mL

(NH4)2SO4 in H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013842.AP
also known as 013842-AP
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Specifications
Chemical Name or MaterialSulfur, plasma standard solution
Concentration or Composition (by Analyte or Components)Ammonium sulfate: 0.413%; Water: 99.587%
MDL NumberMFCD00085316
Molecular Formula(NH4)2SO4 in H2O
Physical FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only