Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
Thermo Scientific Chemicals

Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL

CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013852.AE
also known as 013852-AE
Price (USD)
72.65
Online Exclusive
80.40
Save 7.75 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
72.65
Online Exclusive
80.40
Save 7.75 (10%)
Each
Specifications
CAS471-34-1
CAS7732-18-5
ChEBICHEBI:3311
Chemical Name or MaterialCalcium
ColorColorless
View more
Calcium, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only