Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL
Thermo Scientific Chemicals

Aluminum, plasma standard solution, Specpure™ Al 1000μg/mL

CAS: 7429-90-5 | Al | 26.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 013856.AD
also known as 013856-AD
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.30
Save 8.65 (15%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
56.30
Save 8.65 (15%)
Each
Specifications
CAS7429-90-5
CAS7732-18-5
ChEBICHEBI:33629
Chemical Name or MaterialAluminum
Concentration or Composition (by Analyte or Components)Hydrochloric acid: 5.0%; Aluminum: 0.1%; Water: 94.9%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only