Specifications
Chemical Name or MaterialBoron plasma standard solution
Health Hazard 1H315-H319-H360FD
Health Hazard 2GHS H Statement
H360-H315-H319
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
MDL NumberMFCD00134034
View more