Boron, plasma standard solution, Specpure™ B 1000μg/mL
Thermo Scientific Chemicals

Boron, plasma standard solution, Specpure™ B 1000μg/mL

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013859.AE
also known as 013859-AE
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.70
Save 11.05 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
74.70
Save 11.05 (15%)
Each
Specifications
Chemical Name or MaterialBoron plasma standard solution
Health Hazard 1H315-H319-H360FD
Health Hazard 2GHS H Statement
H360-H315-H319
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P201-P202-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P501c
MDL NumberMFCD00134034
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only