Rubidium, plasma standard solution, Specpure™ Rb 1000μg/mL
Rubidium, plasma standard solution, Specpure™ Rb 1000μg/mL
Rubidium, plasma standard solution, Specpure™ Rb 1000μg/mL
Thermo Scientific Chemicals

Rubidium, plasma standard solution, Specpure™ Rb 1000μg/mL

CAS: 584-09-8 | CO3Rb2 | 230.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
Catalog number 013872.AD
also known as 013872-AD
Price (USD)
54.00
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
54.00
Each
Specifications
CAS584-09-8
CAS7697-37-2
CAS7732-18-5
Chemical Name or MaterialRubidium
ColorColorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only