Strontium, plasma standard solution, Specpure™ Sr 1000μg/mL
Thermo Scientific Chemicals

Strontium, plasma standard solution, Specpure™ Sr 1000μg/mL

Sr(NO3)2 in 5% HNO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013874.AD
also known as 013874-AD
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.20
Save 8.55 (15%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
48.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.20
Save 8.55 (15%)
Each
Specifications
Chemical Name or MaterialStrontium, plasma standard solution
Concentration or Composition (by Analyte or Components)Nitric acid: 5%; Strontium nitrate: 0.24%; Water: 94.76%
DOT InformationDOT Class: 8; Packaging Group: III
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only