Specifications
Chemical Name or MaterialStrontium, plasma standard solution
Concentration or Composition (by Analyte or Components)Nitric acid: 5%; Strontium nitrate: 0.24%; Water: 94.76%
DOT InformationDOT Class: 8; Packaging Group: III
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
View more