Praseodymium foil, 0.1mm (0.004in) thick, 99% (REO)
Praseodymium foil, 0.1mm (0.004in) thick, 99% (REO)
Praseodymium foil, 0.1mm (0.004in) thick, 99% (REO)
Thermo Scientific Chemicals

Praseodymium foil, 0.1mm (0.004in) thick, 99% (REO)

CAS: 7440-10-0 | Pr | 140.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 013962.FF
also known as 013962-FF
Price (USD)
322.65
Online Exclusive
358.00
Save 35.35 (10%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
322.65
Online Exclusive
358.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS7440-10-0
IUPAC Namepraseodymium
Molecular FormulaPr
InChI KeyPUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES[Pr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Praseodymium optical properties for use in amplification of telecommunication systems, including as a doping agent in fluoride fibers. It is also used in the scintillator for medical CAT scans. Used as additive for special steel, non-ferric alloy, hydrogen storage alloy, also reducer for making other rare Earth metal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
RUO – Research Use Only