Rhenium foil, 0.5mm (0.02in) thick, annealed, 99.97% (metals basis)
Rhenium foil, 0.5mm (0.02in) thick, annealed, 99.97% (metals basis)
Rhenium foil, 0.5mm (0.02in) thick, annealed, 99.97% (metals basis)
Thermo Scientific Chemicals

Rhenium foil, 0.5mm (0.02in) thick, annealed, 99.97% (metals basis)

CAS: 7440-15-5 | Re | 186.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 013983.FF
also known as 013983-FF
Price (USD)
822.65
Online Exclusive
914.00
Save 91.35 (10%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
822.65
Online Exclusive
914.00
Save 91.35 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-15-5
ChEBICHEBI:49882
Chemical Name or MaterialRhenium foil
EINECS Number231-124-5
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only