Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis)
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis)
Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis)
Thermo Scientific Chemicals

Vanadium rod, 6.35mm (0.25in) dia, 99.5% (metals basis)

CAS: 7440-62-2 | V | 50.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 cm
25 cm
100 cm
Catalog number 013996.BY
also known as 013996-BY
Price (USD)
1,953.65
Online Exclusive
2,171.00
Save 217.35 (10%)
Each
Quantity:
100 cm
Request bulk or custom format
Price (USD)
1,953.65
Online Exclusive
2,171.00
Save 217.35 (10%)
Each
Chemical Identifiers
CAS7440-62-2
IUPAC Namevanadium
Molecular FormulaV
InChI KeyLEONUFNNVUYDNQ-UHFFFAOYSA-N
SMILES[V]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentDiameter: 6.35 mm
Trace Metal≤0.5% (≤5000ppm)
Appearance (Color)Silver
FormRod
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only