Aluminum isopropoxide, 98+%
Aluminum isopropoxide, 98+%
Aluminum isopropoxide, 98+%
Thermo Scientific Chemicals

Aluminum isopropoxide, 98+%

Reagent for the reduction of carbonyl compounds | CAS: 555-31-7 | C9H21AlO3 | 204.246 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number 014007.A7
also known as 014007-A7
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Quantity:
5 kg
Request bulk or custom format
Price (USD)
209.65
Online Exclusive
233.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS555-31-7
IUPAC Namealuminium(3+) tris(propan-2-olate)
Molecular FormulaC9H21AlO3
InChI KeySMZOGRDCAXLAAR-UHFFFAOYSA-N
SMILES[Al+3].CC(C)[O-].CC(C)[O-].CC(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormGranules and Powder
Assay (unspecified)> 98.0%
Reagent for the reduction of carbonyl compounds. Used in alcoholysis and ester exchange reactions. Also used in synthesis of higher alkoxides, chelates, and acrylates. Aluminum isopropoxide is a useful reagent in the synthesis of alkoxides, chelates and acrylates. It acts as an intermediate of pharmaceutical products like isophytol, testosterone, progesterone and ethisterone. Also used as the material of aluminate coupling agent. It is also used in alcoholysis and ester exchange reactions. In Meerwein-Ponndorf-Verley (MPV) reduction, it is used as a catalyst for the reduction of ketones and aldehydes to their corresponding alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Reagent for the reduction of carbonyl compounds
  • Used in alcoholysis and ester exchange reactions
  • Also used in synthesis of higher alkoxides, chelates, and acrylates
RUO – Research Use Only