Germanium(IV) iodide, 99.999% (metals basis)
Germanium(IV) iodide, 99.999% (metals basis)
Germanium(IV) iodide, 99.999% (metals basis)
Thermo Scientific Chemicals

Germanium(IV) iodide, 99.999% (metals basis)

CAS: 13450-95-8 | GeI4 | 580.248 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 014009.09
also known as 014009-09
Price (USD)
270.00
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
270.00
Each
Chemical Identifiers
CAS13450-95-8
Molecular FormulaGeH8I4
InChI KeyWTPFKACBVKJUIS-UHFFFAOYSA-N
SMILES[GeH4].I.I.I.I
Molecular Weight (g/mol)588.31
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Germanium(IV) iodide is a biochemical for proteomics research. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only