Potassium tin(IV) oxide trihydrate, 95%
Potassium tin(IV) oxide trihydrate, 95%
Potassium tin(IV) oxide trihydrate, 95%
Thermo Scientific Chemicals

Potassium tin(IV) oxide trihydrate, 95%

CAS: 12125-03-0 | H6K2O6Sn | 298.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 014015.30
also known as 014015-30
Price (USD)
44.30
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
44.30
Each
Chemical Identifiers
CAS12125-03-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white
FormPowder
Assay from Supplier's CofATin (Sn): ≥37.0%
Potassium tin(IV) oxide is a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only