Manganese phosphide, 99% (metals basis)
Manganese phosphide, 99% (metals basis)
Manganese phosphide, 99% (metals basis)
Manganese phosphide, 99% (metals basis)
Thermo Scientific Chemicals

Manganese phosphide, 99% (metals basis)

CAS: 12263-33-1 | Mn3P2 | 226.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 014020.09
also known as 014020-09
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
113.65
Online Exclusive
126.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS12263-33-1
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Manganese phosphide is used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only