Di-n-butyltin oxide
Di-n-butyltin oxide
Di-n-butyltin oxide
Thermo Scientific Chemicals

Di-n-butyltin oxide

CAS: 818-08-6 | C8H18OSn | 248.941 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014117.18
also known as 014117-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Specifications
Beilstein4126243
CAS818-08-6
Chemical Name or MaterialDi-n-butyltin oxide
ColorWhite
DOT InformationTransport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
View more
Di-n-butyltin oxide is used as a reagent and a catalyst. It is particularly useful in regioselective alkylation, acylation and sulfonation reactions for starting materials containing alcohol functional groups. It is used in studies pertaining to discovering novel antitumor agents, antifungal activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only