Iron wire, 1.2mm (0.047in) dia, 99+% (metals basis), Thermo Scientific Chemicals
Iron wire, 1.2mm (0.047in) dia, 99+% (metals basis), Thermo Scientific Chemicals
Iron wire, 1.2mm (0.047in) dia, 99+% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iron wire, 1.2mm (0.047in) dia, 99+% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
014147.H4
also known as 014147-H4
100 m
Catalog number 014147.H4
also known as 014147-H4
Price (USD)
726.65
Special offer
Online exclusive
Ends: 30-Jun-2026
855.00
Save 128.35 (15%)
Each
Quantity:
100 m
Request bulk or custom format
Price (USD)
726.65
Special offer
Online exclusive
Ends: 30-Jun-2026
855.00
Save 128.35 (15%)
Each
Chemical Identifiers
CAS7439-89-6
IUPAC Nameiron
Molecular FormulaFe
InChI KeyXEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES[Fe]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only