Tantalum(V) chloride, 99.8% (metals basis)
Tantalum(V) chloride, 99.8% (metals basis)
Tantalum(V) chloride, 99.8% (metals basis)
Tantalum(V) chloride, 99.8% (metals basis)
Thermo Scientific Chemicals

Tantalum(V) chloride, 99.8% (metals basis)

CAS: 7721-01-9 | Cl5Ta | 358.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 014164.06
also known as 014164-06
Price (USD)
50.65
Online exclusive
55.80
Save 5.15
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
50.65
Online exclusive
55.80
Save 5.15
Each
Chemical Identifiers
CAS7721-01-9
IUPAC Nametantalum(5+) pentachloride
Molecular FormulaCl5Ta
InChI KeyOEIMLTQPLAGXMX-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder or crystalline powder or crystals
Total Metal Impurities≤0.2% (2000 ppm)
In chlorination of organic substances, as an intermediate, and in the production of pure metalTantalum(V) chloride is used in the preparation of catalyst for the polycyclotrimerizations of alkenediynes, chloro-aryloxide compounds and for the plasma-enhanced atomic layer deposition of tantalum nitride films. It acts as a starting material for new edge-bridged octahedral M6 cluster compounds. It is involved in the preparation of tantalum(V) oxychloride, tantalum pentoxide. It is electrophilic and behaves like a Friedel-Crafts type catalyst, similar to aluminum(III) chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In chlorination of organic substances, as an intermediate, and in the production of pure metal
  • Ampouled under argon
RUO – Research Use Only