Copper(II) acetate monohydrate, ACS, 98.0-102.0%
Copper(II) acetate monohydrate, ACS, 98.0-102.0%
Copper(II) acetate monohydrate, ACS, 98.0-102.0%
Thermo Scientific Chemicals

Copper(II) acetate monohydrate, ACS, 98.0-102.0%

CAS: 6046-93-1 | C4H8CuO5 | 199.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 014177.22
also known as 014177-22
Price (USD)
53.65
Online Exclusive
58.90
Save 5.25 (9%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
58.90
Save 5.25 (9%)
Each
Chemical Identifiers
CAS6046-93-1
IUPAC Namecopper(2+) diacetate hydrate
Molecular FormulaC4H8CuO5
InChI KeyNWFNSTOSIVLCJA-UHFFFAOYSA-L
SMILESO.[Cu++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCa 0.005%
Impurity contentFe 0.002%
Impurity contentNa 0.05%
Insoluble material0.01%
Impurity contentK 0.01%
View more
Copper(II) acetate monohydrate is used in DNA extraction in biochemical applications. It is also used as a catalyst or oxidizing agent in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only