Nickel plate, 6.35mm (0.25in) thick, annealed, 99.5% (metals basis)
Nickel plate, 6.35mm (0.25in) thick, annealed, 99.5% (metals basis)
Nickel plate, 6.35mm (0.25in) thick, annealed, 99.5% (metals basis)
Thermo Scientific Chemicals

Nickel plate, 6.35mm (0.25in) thick, annealed, 99.5% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
Catalog number 014194.GH
also known as 014194-GH
Price (USD)
425.65
Online Exclusive
473.00
Save 47.35 (10%)
Each
Quantity:
100 x 100 mm
Request bulk or custom format
Price (USD)
425.65
Online Exclusive
473.00
Save 47.35 (10%)
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.5% (metals basis)
Appearance (Color)Silvery
FormPlate, 6.35mm (0.25in) thick, annealed.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only