Strontium, Oil based standard solution, Specpure™ Sr 1000μg/g
Thermo Scientific Chemicals

Strontium, Oil based standard solution, Specpure™ Sr 1000μg/g

 
Have Questions?
Catalog NumberQuantity
014205.18
also known as 014205-18
50 g
Catalog number 014205.18
also known as 014205-18
Price (USD)
81.65
Online Exclusive
91.10
Save 9.45 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
81.65
Online Exclusive
91.10
Save 9.45 (10%)
Each
Specifications
Chemical Name or MaterialStrontium Oil based standard solution
MDL NumberMFCD00134060
Name NoteSr 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only