Specifications
Chemical Name or MaterialLead Oil based standard solution
Health Hazard 1H304-H350-H360
Health Hazard 2GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P201-P202-P281-P301+P310-P308+P313-P331-P501c
MDL NumberMFCD00134050
View more