Chromium, Oil based standard solution, Specpure™ Cr 1000μg/g
Thermo Scientific Chemicals

Chromium, Oil based standard solution, Specpure™ Cr 1000μg/g

 
Have Questions?
Catalog NumberQuantity
014222.18
also known as 014222-18
50 g
Catalog number 014222.18
also known as 014222-18
Price (USD)
77.60
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
77.60
Each
Specifications
Chemical Name or MaterialChromium Oil based standard solution
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL NumberMFCD00010944
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only