Lanthanum, Oil based standard solution, Specpure™ La 1000μg/g
Thermo Scientific Chemicals

Lanthanum, Oil based standard solution, Specpure™ La 1000μg/g

 
Have Questions?
Catalog NumberQuantity
014224.18
also known as 014224-18
50 g
Catalog number 014224.18
also known as 014224-18
Price (USD)
114.65
Special offer
Online exclusive
Ends: 30-Jun-2026
135.00
Save 20.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
114.65
Special offer
Online exclusive
Ends: 30-Jun-2026
135.00
Save 20.35 (15%)
Each
Specifications
Chemical Name or MaterialLanthanum Oil based standard solution
MDL NumberMFCD00011066
Name NoteLa 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only