Boron, Oil based standard solution, Specpure™ B 1000μg/g
Thermo Scientific Chemicals

Boron, Oil based standard solution, Specpure™ B 1000μg/g

 
Have Questions?
Catalog NumberQuantity
014227.18
also known as 014227-18
50 g
Catalog number 014227.18
also known as 014227-18
Price (USD)
85.65
Online Exclusive
95.20
Save 9.55 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
85.65
Online Exclusive
95.20
Save 9.55 (10%)
Each
Specifications
Chemical Name or MaterialBoron Oil based standard solution
MDL NumberMFCD00134034
Name NoteB 1000μg/g
Physical FormLiquid
Quantity50 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only