Specifications
Chemical Name or MaterialCadmium Oil based standard solution
Health Hazard 1H304-H350
Health Hazard 2GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3P201-P202-P281-P301+P310-P308+P313-P331-P501c
MDL NumberMFCD00010914
View more