Lead(II,IV) oxide, 97% (metals basis)
Lead(II,IV) oxide, 97% (metals basis)
Lead(II,IV) oxide, 97% (metals basis)
Thermo Scientific Chemicals

Lead(II,IV) oxide, 97% (metals basis)

CAS: 1314-41-6 | O4Pb3 | 685.596 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014232.A1
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS1314-41-6
IUPAC Namebicyclo[3.1.1]triplumboxan-1-yl
Molecular FormulaO4Pb3
InChI KeyXMFOQHDPRMAJNU-UHFFFAOYSA-N
SMILESO1[Pb]2O[Pb]1O[Pb]O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
FormPowder
Assay from Supplier's CofAPb₃O₄: ≥96.0%
Lead(II,IV) oxide is used to prepare colorless glass, faience glaze, porcelain painting flux, iron and steel coatings, rubber pigment and in glass cement. It is also used in gas and steam pipes, storage batteries, writing on glass and to make lead peroxide and matches. It is associated with linseed oil and used as a thick, long-lasting anti-corrosive paint. It gives better water resistant properties by replacing magnesium oxide. Further, it is used in the manufacture of lead glass and rustproof primer paints. In addition to this, it acts as a pigment for primer paints for iron objects.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only