Silver tubing, 6.35mm (0.25in) OD, 5.84mm (0.230in) ID, 99.9% (metals basis)
Silver tubing, 6.35mm (0.25in) OD, 5.84mm (0.230in) ID, 99.9% (metals basis)
Silver tubing, 6.35mm (0.25in) OD, 5.84mm (0.230in) ID, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver tubing, 6.35mm (0.25in) OD, 5.84mm (0.230in) ID, 99.9% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
4 x 50 cm
Catalog number 014252.BW
also known as 014252-BW
Price (USD)
356.00
Each
Quantity:
50 cm
Request bulk or custom format
Price (USD)
356.00
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-22-4
ChEBICHEBI:9141
Chemical Name or MaterialSilver tubing
EINECS Number231-131-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only