Ammonium bromide, ACS, 99.0% min
Ammonium bromide, ACS, 99.0% min
Ammonium bromide, ACS, 99.0% min
Ammonium bromide, ACS, 99.0% min
Thermo Scientific Chemicals

Ammonium bromide, ACS, 99.0% min

CAS: 12124-97-9 | BrH4N | 97.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2 kg
Catalog number 014257.A3
also known as 014257-A3
Price (USD)
172.65
Special offer
Online exclusive
Ends: 30-Jun-2026
203.00
Save 30.35 (15%)
Each
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)
172.65
Special offer
Online exclusive
Ends: 30-Jun-2026
203.00
Save 30.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS12124-97-9
IUPAC Nameammonium bromide
Molecular FormulaBrH4N
InChI KeySWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES[NH4+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormSolid
Assay (unspecified)≤99.0% NH4Br
Barium content≤0.002%
Chloride content≤0.2%
View more
Corrosion inhibitors, manufacturing of photographic films, plates, paper, lithography, fireproofing of woodsAmmonium bromide is often used as an intermediate source of bromide ion in laboratory processes and pharmaceutical preparations. It is most commonly used as an additive in the preparation of photographic films, fireproofing products and corrosion inhibitors. Ammonium bromide is widely utilized in the manufacture of photographic chemicals and emulsions. It is also used in pharmaceutical preparations. It finds application in fireproofing of wood and lithography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only