Tantalum Thinfoil, 0.0127mm (0.0005in) thick, 99.95% (metals basis)
Tantalum Thinfoil, 0.0127mm (0.0005in) thick, 99.95% (metals basis)
Tantalum Thinfoil, 0.0127mm (0.0005in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Tantalum Thinfoil, 0.0127mm (0.0005in) thick, 99.95% (metals basis)

CAS: 7440-25-7 | Ta | 180.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 100 mm
50 x 500 mm
50 x 2000 mm
Catalog number 014266.FY
also known as 014266-FY
Price (USD)
67.65
Online Exclusive
75.20
Save 7.55 (10%)
Each
Quantity:
50 x 100 mm
Request bulk or custom format
Price (USD)
67.65
Online Exclusive
75.20
Save 7.55 (10%)
Each
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.0127mm (0.0005in) thick.
Assay from Supplier's CofA99.95% (metals basis)
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only