Calcium carbide
Calcium carbide
Calcium carbide
Thermo Scientific Chemicals

Calcium carbide

CAS: 75-20-7 | C2Ca
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 x 1 kg
Catalog number 014308.A1
also known as 014308-A1
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS75-20-7
IUPAC Namecalcium acetylene
Molecular FormulaC2Ca
InChI KeyUIXRSLJINYRGFQ-UHFFFAOYSA-N
SMILES[Ca++].[C-]#[C-]
View more
SpecificationsSpecification SheetSpecification Sheet
FormLump
Transition TemperatureGray to black
Assay (unspecified)Ca 46% min
Calcium carbide is used as a desulfurizer, dehydrant of steel, fuel in steel making, powerful deoxidizer and as a source of acetylene gas. It is used as a starting material for the preparation of calcium cyanamide, ethylene, chloroprene rubber, acetic acid, dicyandiamide and cyanide acetate. It is used in carbide lamps, toy cannons such as the big-bang cannon and bamboo cannon. It is associated with calcium phosphide and used in floating, self-igniting naval signal flares. Further, it is involved in the reduction of copper sulfide to metallic copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only