Sodium azide, 99%
Sodium azide, 99%
Sodium azide, 99%
Thermo Scientific Chemicals

Sodium azide, 99%

CAS: 26628-22-8 | N3Na | 65.011 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 014314.36
also known as 014314-36
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS26628-22-8
IUPAC Namesodium azide
Molecular FormulaN3Na
InChI KeyPXIPVTKHYLBLMZ-UHFFFAOYSA-N
SMILES[Na+].[N-]=[N+]=[N-]
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals (as Pb)≤20ppm (0.002%) (non-U.S. specification)
pH≥9 (5% aq. solution) (non-U.S. specification)
Appearance (Color)White to pale cream
FormPowder or crystalline powder
Assay from Suppliers CofA≥98.5%
View more
In airbag inflation, as a preservative in diagnostic medicinals.Sodium azide is used in organic synthesis for the introduction of an azide group to prepare a desired compound. It acts as a precursor to prepare inorganic azides like lead azide and silver azide. In biochemistry, it is useful as a probe reagent and a preservative. It also acts as a nitridizing agent and cytochrome oxidase inhibitor. Further, it is used as a propellant in airbags.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only