Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis)
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis)
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis)
Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis)
Thermo Scientific Chemicals

Nickel wire, 1.0mm (0.04in) dia, annealed, 99.5% (metals basis)

CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 m
50 m
Catalog number 014337.G6
also known as 014337-G6
Price (USD)
75.65
Online Exclusive
83.70
Save 8.05 (10%)
Each
Quantity:
10 m
Request bulk or custom format
Price (USD)
75.65
Online Exclusive
83.70
Save 8.05 (10%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS7440-02-0
ChEBICHEBI:28112
Chemical Name or MaterialNickel wire
EINECS Number231-111-4
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only