Standard Sample Tube, 3.24mm ID
Thermo Scientific Chemicals

Standard Sample Tube, 3.24mm ID

For use with MKII Magnetic Susceptibility Balances
Have Questions?
Change viewbuttonViewtableView
Quantity:
20 Pack
5 Pack
1 Each
Catalog number 014349.KL
also known as 014349-KL
Price (USD)
938.65
Online Exclusive
1,043.00
Save 104.35 (10%)
Each
Quantity:
20 Pack
Request bulk or custom format
Price (USD)
938.65
Online Exclusive
1,043.00
Save 104.35 (10%)
Each
Specifications
Quantity20 Pack
Recommended StorageAmbient temperatures
TypeStandard Sample Tube
Unit SizeEach
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • I.D.: 3.24mm
RUO – Research Use Only