Niobium foil, 0.5mm (0.02in) thick, annealed, 99.8% (metals basis)
Niobium foil, 0.5mm (0.02in) thick, annealed, 99.8% (metals basis)
Niobium foil, 0.5mm (0.02in) thick, annealed, 99.8% (metals basis)
Thermo Scientific Chemicals

Niobium foil, 0.5mm (0.02in) thick, annealed, 99.8% (metals basis)

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 100 mm
100 x 250 mm
100 x 500 mm
Catalog number 014358.FY
also known as 014358-FY
Price (USD)
111.00
Each
Quantity:
50 x 100 mm
Request bulk or custom format
Price (USD)
111.00
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.5mm (0.02in) thick, annealed.
Assay from Supplier's CofA≥99.8% (metals basis)
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only